MilliporeSigma
  • Synthesis of benzimidazoles by PIDA-promoted direct C(sp2)-H imidation of N-arylamidines.
Synthesis of benzimidazoles by PIDA-promoted direct C(sp2)-H imidation of N-arylamidines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-09-22)
Jinbo Huang, Yimiao He, Yong Wang, Qiang Zhu
ABSTRACT
A metal-free synthesis of diversified benzimidazoles from N-arylamidines through a phenyliodine(III) diacetate (PIDA) promoted intramolecular direct C(sp(2))-H imidation has been developed. The reaction proceeds smoothly at 0 °C or ambient temperature to provide the desired products in good to excellent yields. The synthesis of 2-alkyl- or 2-alkyl-fused benzimidazoles, which are generally inaccessible by similar Pd- or Cu-catalyzed approaches, can also be achieved.
Sigma-Aldrich
(Diacetoxyiodo)benzene, 98%