MilliporeSigma
  • A rhodium complex-linked β-barrel protein as a hybrid biocatalyst for phenylacetylene polymerization.
A rhodium complex-linked β-barrel protein as a hybrid biocatalyst for phenylacetylene polymerization.
Chemical communications (Cambridge, England) (2012-08-25)
Akira Onoda, Kazuki Fukumoto, Marcus Arlt, Marco Bocola, Ulrich Schwaneberg, Takashi Hayashi
ABSTRACT
Our group recently prepared a hybrid catalyst containing a rhodium complex, Rh(Cp)(cod), with a maleimide moiety at the peripheral position of the Cp ligand. This compound was then inserted into a β-barrel protein scaffold of a mutant of aponitrobindin (Q96C) via a covalent linkage. The hybrid protein is found to act as a polymerization catalyst and preferentially yields trans-poly(phenylacetylene) (PPA), although the rhodium complex without the protein scaffold normally produces cis PPA.
Sigma-Aldrich
Phenylacetylene, 98%