MilliporeSigma
  • Tautomers of anthrahydroquinones: enzymatic reduction and implications for chrysophanol, monodictyphenone, and related xanthone biosyntheses.
Tautomers of anthrahydroquinones: enzymatic reduction and implications for chrysophanol, monodictyphenone, and related xanthone biosyntheses.
Journal of the American Chemical Society (2012-08-23)
Michael A Schätzle, Syed Masood Husain, Sascha Ferlaino, Michael Müller
ABSTRACT
Reduction of emodin by sodium dithionite resulted in the formation of two tautomeric forms of emodin hydroquinone. Subsequent conversion by the short-chain dehydrogenase/reductase (SDR) MdpC into the corresponding 3-hydroxy-3,4-dihydroanthracen-1(2H)-one implies that deoxygenation is the first step in monodictyphenone biosynthesis. Implications for chrysophanol formation as well as reaction sequences in the related xanthone, ergochrome, and bianthraquinone biosyntheses are discussed.
Sigma-Aldrich
Xanthone, 97%