MilliporeSigma
  • Synthesis of high-quality diesel with furfural and 2-methylfuran from hemicellulose.
Synthesis of high-quality diesel with furfural and 2-methylfuran from hemicellulose.
ChemSusChem (2012-08-22)
Guangyi Li, Ning Li, Zhiqiang Wang, Changzhi Li, Aiqin Wang, Xiaodong Wang, Yu Cong, Tao Zhang
ABSTRACT
Hydroxyalkylation-alkylation (HAA) coupled with hydrodeoxygenation is a promising route for the synthesis of renewable high-quality diesel or jet fuel. In this work, a series of solid-acid catalysts were firstly used for HAA between lignocellulose-derived furan and carbonyl compounds. Among the investigated catalysts, Nafion-212 resin demonstrated the highest activity and stability. Owing to the high activity of the reactants and the advantage in industrial integration, the HAA of 2-methylfuran (2-MF) and furfural can be considered as a prospective route in future applications. Catalyst loading, reaction temperature, and time had evident effects on the HAA of 2-MF and furfural over Nafion-212 resin. Finally, the HAA product of 2-MF and furfural was hydrogenated over a Pd/C catalyst and hydrodeoxygenated over Pt-loaded solid-acid catalysts. Pt/zirconium phosphate (Pt/ZrP) was found to be the best catalyst for hydrodeoxygenation. Over the 4 % Pt/ZrP catalyst, a 94 % carbon yield of diesel and 75 % carbon yield of C15 hydrocarbons (with 6-butylundecane as the major component) was achieved.
Sigma-Aldrich
2-Methylfuran, contains 200-400 BHT as stabilizer, 99%
Pricing and availability is not currently available.