MilliporeSigma
  • Reactive oxygen species and the MEK/ERK pathway are involved in the toxicity of clostridium perfringens α-toxin, a prototype bacterial phospholipase C.
Reactive oxygen species and the MEK/ERK pathway are involved in the toxicity of clostridium perfringens α-toxin, a prototype bacterial phospholipase C.
The Journal of infectious diseases (2012-08-21)
Laura Monturiol-Gross, Marietta Flores-Díaz, Cindy Araya-Castillo, María-José Pineda-Padilla, Graeme C Clark, Richard W Titball, Alberto Alape-Girón
ABSTRACT
Clostridium perfringens, the most broadly distributed pathogen in nature, produces a prototype phospholipase C, also called α-toxin, which plays a key role in the pathogenesis of gas gangrene. α-Toxin causes plasma membrane disruption at high concentrations, but the role of intracellular mediators in its toxicity at low concentrations is unknown. This work demonstrates that α-toxin causes oxidative stress and activates the MEK/ERK pathway in cultured cells and furthermore provides compelling evidence that O(2)(-.), hydrogen peroxide, and the OH(.) radical are involved in its cytotoxic and myotoxic effects. The data show that antioxidants and MEK1 inhibitors reduce the cytotoxic and myotoxic effects of α-toxin and demonstrate that edaravone, a clinically used hydroxyl radical trap, reduces the myonecrosis and the mortality caused by an experimental infection with C. perfringens in a murine model of gas gangrene. This knowledge provides new insights for the development of novel therapies to reduce tissue damage during clostridial myonecrosis.
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%