MilliporeSigma
  • Studies on chemical structure modification and structure-activity relationship of derivatives of gambogic acid at C(39).
Studies on chemical structure modification and structure-activity relationship of derivatives of gambogic acid at C(39).
Chemistry & biodiversity (2012-08-18)
Hao-Peng Sun, Zong-Liang Liu, Xin Xue, Yuan Gao, Lei Zhang, Jin-Xin Wang, Qing-Long Guo, Qi-Dong You
ABSTRACT
The natural product gambogic acid exhibits high potency in inhibiting cancer cell lines. Rational medicinal modifications on gambogic acid will improve its physicochemical properties and drug-like characters. To investigate the structure-activity relationship of gambogic acid and also to find rational modification position on its chemical skeleton, we designed, synthesized, and characterized 16 derivatives of gambogic acid that were modified at C(39). The structure-activity relationships (SARs) were discussed. The anti-proliferation data were accquired through MTT (=3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-2H-tetrazolium bromide) assays of A549, BGC823, U251, HepG2, and MDA-MB-231 cancer cell lines. Most of the synthesized compounds showed strong inhibitory effects. The SAR study revealed that derivatives with aliphatic amino moieties at C(39) were more potent than those with other substituents. The C(39) position can undergo different kinds of chemical modifications without leading to loss of activity. Compounds 4 and 6 can serve as potential lead compounds for further development of new anticancer drugs.