MilliporeSigma
  • Vasospastic angina with J-wave pattern and polymorphic ventricular tachycardia effectively treated with quinidine.
Vasospastic angina with J-wave pattern and polymorphic ventricular tachycardia effectively treated with quinidine.
Annals of noninvasive electrocardiology : the official journal of the International Society for Holter and Noninvasive Electrocardiology, Inc (2012-07-24)
Jerzy Sacha, Szymon Barabach, Piotr Feusette, Piotr Kukla
ABSTRACT
Myocardial ischemia during coronary spasm may generate malignant ventricular arrhythmias. The J-wave pattern was suggested to be a marker of a disorder associated with life-threatening arrhythmias. We report the case of a patient with vasospastic angina and J-wave pattern in inferior and lateral leads associated with polymorphic ventricular tachycardia which was effectively treated only with quinidine-vasodilating drugs were not able to prevent the arrhythmia although they were effective in preventing ischemic events. The J-wave pattern in inferolateral leads may be a sign of electrical vulnerability to lethal ventricular arrhythmia in patients suffering from vasospastic angina--quinidine can effectively prevent such arrhythmias in these patients.
Sigma-Aldrich
Dianhydro-D-glucitol, 98%
Pricing and availability is not currently available.