MilliporeSigma
  • Effects of dosing interval on the pharmacokinetic and pharmacodynamic interactions between the oral adsorbent AST-120 and triazolam in humans.
Effects of dosing interval on the pharmacokinetic and pharmacodynamic interactions between the oral adsorbent AST-120 and triazolam in humans.
European journal of clinical pharmacology (2012-05-31)
Tsutomu Kotegawa, Kimiko Tsutsumi, Hajime Morita, Hiromistu Imai, Misaki Morita, Tsuneaki Yoshizato, Tetsuji Ohyama, Shinya Uchida, Hiroshi Watanabe, Noriyuki Namiki, Kyoichi Ohashi
ABSTRACT
The objective of this study was to evaluate the pharmacokinetic and pharmacodynamic interactions between the oral adsorbent AST-120 and triazolam. In this randomized, cross-over study, 12 healthy volunteers received a single oral dose of triazolam 0.25 mg alone or with AST-120 2 g given 0, 30 or 60 min before triazolam administration. The area under the plasma triazolam concentration-time curve (AUC(0-∞)) significantly decreased with simultaneous AST-120 + triazolam (alone vs simultaneous: 10.9 ± 6.0 vs 6.4 ± 2.6 ng·h/mL, p = 0.003). Triazolam-induced impairment in psychomotor performance assessed by the digit symbol substitution test was significantly attenuated when AST-120 was administered simultaneously. No significant changes in pharmacokinetic and pharmacodynamic parameters were observed when AST-120 was given 30 or 60 min before triazolam administration. Administering AST-120 simultaneously with triazolam affects the pharmacokinetics and pharmacodynamics of triazolam. Dosing AST-120 at least 30 min before triazolam administration may avoid these interactions.
Product Description
Supelco
Triazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®