MilliporeSigma
  • Protein quantification methods to determine protein concentration prior to electrophoresis.
Protein quantification methods to determine protein concentration prior to electrophoresis.
Methods in molecular biology (Clifton, N.J.) (2012-05-16)
J P Dean Goldring
ABSTRACT
During each step of a protein isolation technique, if enzyme activity is to be determined and before a protein mixture is separated on a polyacrylamide electrophoresis gel, it is important to determine the concentration of the protein(s) in solution. Measuring protein concentration involves absorbance in the UV range or staining the protein with dyes or copper. This review describes the various protein determination methods that can be employed to measure protein concentration in solution.
Sigma-Aldrich
2,2′-Biquinoline-4,4′-dicarboxylic acid, ≥90% (TLC)