MilliporeSigma
  • New glycosidic alkaloid from the nearly ripe fruits of Euodia rutaecarpa.
New glycosidic alkaloid from the nearly ripe fruits of Euodia rutaecarpa.
Journal of Asian natural products research (2012-05-16)
Chuan-Qin Hu, Ke-Ke Li, Xiu-Wei Yang
ABSTRACT
A new natural product, 10-hydroxyrutaecarpine (1), and a rarely new glycosidic alkaloid, rutaecarpine-10-O-rutinoside (2), along with the known compounds rutaecarpine (3), evodiamine, wuzhuyuamide-I, and dehydroevodiamine were isolated from the butanol fraction of 70% ethanol aqueous extract of the dried and nearly ripe fruits of Euodia rutaecarpa (Juss.) Benth. Their structures were elucidated on the basis of their spectroscopic data.
Sigma-Aldrich
Rutaecarpine, >98% (HPLC)