MilliporeSigma
  • Chiral aminophosphines as catalysts for enantioselective double-Michael indoline syntheses.
Chiral aminophosphines as catalysts for enantioselective double-Michael indoline syntheses.
Molecules (Basel, Switzerland) (2012-05-15)
San N Khong, Ohyun Kwon
ABSTRACT
The bisphosphine-catalyzed double-Michael addition of dinucleophiles to electron-deficient acetylenes is an efficient process for the synthesis of many nitrogen-containing heterocycles. Because the resulting heterocycles contain at least one stereogenic center, this double-Michael reaction would be even more useful if an asymmetric variant of the reaction were to be developed. Aminophosphines can also facilitate the double-Michael reaction and chiral amines are more readily available in Nature and synthetically; therefore, in this study we prepared several new chiral aminophosphines. When employed in the asymmetric double-Michael reaction between ortho-tosylamidophenyl malonate and 3-butyn-2-one, the chiral aminophosphines produced indolines in excellent yields with moderate asymmetric induction.
Sigma-Aldrich
3-Butyn-2-one, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Indoline, ReagentPlus®, 99%
Pricing and availability is not currently available.