MilliporeSigma
  • Distribution of antifouling biocides and perfluoroalkyl compounds in sediments from selected locations in Indonesian coastal waters.
Distribution of antifouling biocides and perfluoroalkyl compounds in sediments from selected locations in Indonesian coastal waters.
Archives of environmental contamination and toxicology (2012-05-10)
Hiroya Harino, Zainal Arifin, Inneke F M Rumengan, Takaomi Arai, Madoka Ohji, Nobuyuki Miyazaki
ABSTRACT
Coastal marine environments are considered to be the most sensitive areas for the accumulation of organotin (OT) compounds and other emerging new pollutants, such as perfluoroalkyl compounds. Contamination by these compounds is a matter of great concern due to their accumulation and possible negative impact on the coastal environment and organisms. The concentrations of tributyltin (TBT) compounds were greater in Indonesia, i.e., on the order of Bitung > Manado > Jakarta Bay > Gangga Island, and TBT in sediment from Bitung and Manado was the dominant species among butyltin (BT) compounds. Sea Nine 211, diuron, and irgarol 1051 were detected among alternative biocides in Bitung, Manado, and Gangga Island and irgarol 1051 was detected in Jakarta Bay. Perfluorooctanoic acid (PFOA) and perfluorosulfonic acid (PFOS) in Jakarta Bay were detected at 0.25 to 6.1 μg kg(-1) dry weight (dw) and 0.58 to 3.7 μg kg(-1) dw, respectively, and the concentrations of PFOS at most sampling sites were greater than those of PFOA. Thus, coastal waters from Indonesia have already been contaminated by antifouling biocides and perfluoroalkyl compounds.
Product Description
Supelco
Irgarol®, PESTANAL®, analytical standard