MilliporeSigma
  • Biphasic effects of H. pylori infection on low-dose aspirin-induced gastropathy depending on the gastric acid secretion level.
Biphasic effects of H. pylori infection on low-dose aspirin-induced gastropathy depending on the gastric acid secretion level.
Journal of gastroenterology (2012-05-03)
Katsunori Iijima, Nobuyuki Ara, Yasuhiko Abe, Tomoyuki Koike, Toshimitsu Iwabuchi, Hirohiko Shinkai, Kaname Uno, Hiroyuki Endo, Naoki Asano, Tooru Shimosegawa
ABSTRACT
The association of Helicobacter pylori infection with aspirin-induced gastropathy is controversial. H. pylori infection exerts diverse effects on gastric acid secretion. In this study, the interaction between H. pylori infection and aspirin was investigated with reference to the individual gastric acid secretion level in H. pylori-positive subjects. Ninety-three (81 men, mean age: 70 years) long-term low-dose aspirin takers were prospectively enrolled. H. pylori infection was evaluated by serum IgG antibody determination, and gastrin-stimulated acid output was assessed with the endoscopic gastrin test. H. pylori-positive aspirin-takers were classified into 2 subgroups (hyposecretors and non-hyposecretors). The grade of gastric mucosal injury was assessed endoscopically according to the modified Lanza score; intensive aspirin-induced gastropathy was defined as a modified Lanza score of ≥4. Multiple logistic regression analyses were used to adjust for potential confounders. With H. pylori-negative patients taken as the reference, H. pylori infection was found to be positively associated with intensive gastropathy among non-hyposecretors, with an odds ratio (OR) (95 % confidence interval [CI]) of 4.2 (1.1-17.1), while the infection was negatively associated with gastropathy among hyposecretors, with an OR (95 % CI) of 0.3 (0.08-0.9). Aspirin-induced gastropathy occurred preferentially in the antrum among H. pylori-positive non-hyposecretors, while it affected the fundus among H. pylori-positive hyposecretors. The effect of H. pylori infection on the aspirin-induced gastropathy was biphasic depending on the individual gastric acid secretion level. In the presence of sufficient amounts of gastric acid, H. pylori infection and aspirin could synergistically damage gastric mucosal integrity, while in the absence of sufficient amounts of gastric acid, the synergistic effect could be completely counteracted and the infection could even suppress the aspirin-induced gastropathy.
Sigma-Aldrich
Pentagastrin, ≥95% (HPLC), powder