MilliporeSigma
  • [Uptake of 3-methyl-3-buten-1-ol into aqueous mixed solution of sulfuric acid and hydrogen peroxide].
[Uptake of 3-methyl-3-buten-1-ol into aqueous mixed solution of sulfuric acid and hydrogen peroxide].
Huan jing ke xue= Huanjing kexue (2012-04-04)
Tian-He Wang, Ze Liu, Mao-Fa Ge, Wei-Gang Wang
ABSTRACT
Multiphase acid-catalyzed oxidation with hydrogen peroxide (H2O2) has been suggested recently to be a potential route to SOA formation, but the kinetics and chemical mechanism of this process have not been well-known yet. In this work, the uptake of 3-methyl-3-buten-1-ol (MBO331) into aqueous mixed solutions of H2O2, and sulfuric acid (H2SO4) was performed using a rotated wetted-wall reactor coupled to a VUV single-photon ionization time of flight mass spectrometer (VUV-SPI-TOFMS). The reactive uptake coefficients (gamma) were acquired for the first time and the reaction pathways were deduced according to products information. The uptake of MBO331 into H2SO4/H2O2 was fast, resulting in gamma reaching 2.52 x 10(-4)-1.05 x 10(-2) for 40%-60% H2SO4. Acetaldehyde, acetone and 3-methyl-3, 4-expoxybutane-1-ol were suggested as gas-phase products in this process. 3-methyl-3,4-expoxybutane-1-ol can transform into polyhydroxy compounds while the further reactions of the carbonyl products can occur in acidic solution, which may play a role in SOA formation. Thus, the heterogeneous acid-catalyzed oxidation of MBO331 with H2O2 might be a significant contributor to SOA loading.
Sigma-Aldrich
3-Methyl-3-buten-1-ol, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methyl-3-buten-2-ol, 98%
Pricing and availability is not currently available.