MilliporeSigma
  • Improving functionality of whole egg powder by the addition of gelatine, lactose, and pullulan.
Improving functionality of whole egg powder by the addition of gelatine, lactose, and pullulan.
Journal of food science (2012-03-16)
Mehmet Koç, Banu Koç, Gonca Susyal, Melike Sakin Yilmazer, Neriman Bağdatlıoğlu, Figen Kaymak-Ertekin
ABSTRACT
The addition of gelatine (G), lactose (L), pullulan (P), and their mixtures at equal ratios (gelatine + lactose [GL] and gelatin + pullulan [GP]) to whole egg prior to drying was studied. Their effects on the functional properties (emulsion and gelling ability, water holding capacity of gel, foaming, color) and soluble protein content of spray dried egg powder during the 6 mo of storage at 20 °C and 50% relative humidity were investigated. It was demonstrated that the emulsion and foaming stability, water holding capacity of gel, and color change were significantly affected by the storage time, whereas storage time did not affect the strength of gel texture prepared by egg powders. Gelatine and pullulan improved the foaming stability and water holding capacity. Lactose caused a decrease in emulsion and foaming stability values. The maximum color change was observed for the plain egg powder, showing that mixing whole egg with carbohydrate- and/or protein-based additives before the drying process preserved the color of egg powder. Adding carbohydrate and/or protein caused significant changes in functional properties of egg powder.
Product Description
Supelco
Pullulan Standard Set Mp ~350 - 700′000, set of analytical standards, for GPC
Sigma-Aldrich
Pullulan, suitable for substrate for pullulanase