MilliporeSigma
  • Synthesis, characterization and biological studies of new antimony(III) halide complexes with ω-thiocaprolactam.
Synthesis, characterization and biological studies of new antimony(III) halide complexes with ω-thiocaprolactam.
Journal of inorganic biochemistry (2012-03-02)
Ibrahim I Ozturk, Christina N Banti, Manos J Manos, Anastasios J Tasiopoulos, Nikolaos Kourkoumelis, Konstantinos Charalabopoulos, Sotiris K Hadjikakou
ABSTRACT
Three new antimony(III) halide complexes (SbX(3), X=Cl, Br and I) with the heterocyclic thione ω-thiocaprolactam (1-azacycloheptane-2-thione, (Hthcl)) of formulae {[SbCl(2)(μ(2)-Cl)(Hthcl)(2)](n)} (1), {[(SbBr(2)(μ(2)-Br)(Hthcl)(2))(2)]} (2) and {[(SbI(2)(μ(2)-I)(Hthcl)(2))(2)]} (3) were synthesized from the reaction of antimony(III) halides with ω-thiocaprolactam in 1:2 stoichiometry. The complexes were characterized by elemental analysis, FT-IR spectroscopy, (1)H, (13)C NMR spectroscopy and Thermal Gravimetry-Differential Thermal Analysis (TG-DTA). Crystal structures of the ligand ω-thiocaprolactam and its complexes 1-3 were determined with single crystal X-ray diffraction analysis. Complexes 1-3 and ω-thiocaprolactam were evaluated for their in vitro cytotoxic activity against leiomyosarcoma (LMS) and human breast adenocarcinoma (MCF-7) tumor cell lines. Antimony complexes 1-3 exhibit strong antiproliferative activity against both cell lines tested. The higher such activity was found for 3 with IC(50) values of 0.12±0.04 μM (LMS) and 0.76±0.16 μM (MCF-7) which are 60 and 10 times respectively, stronger than that of cisplatin. The influence of these complexes 1-3 and ω-thiocaprolactam upon the catalytic peroxidation of linoleic acid to hyperoxolinoleic acid by the enzyme lipoxygenase (LOX) was kinetically and theoretically studied. The results were shown negligible inhibitory activity of 1-3 against LOX.
Sigma-Aldrich
ε-Caprolactam, 99%