MilliporeSigma
  • Displacement immunoassay for the detection of ochratoxin A using ochratoxin B modified glass beads.
Displacement immunoassay for the detection of ochratoxin A using ochratoxin B modified glass beads.
Analytical and bioanalytical chemistry (2012-02-15)
Vasilica Lates, Cheng Yang, Ionel Catalin Popescu, Jean-Louis Marty
ABSTRACT
We report here the development of a new assay for the detection of ochratoxin A (OTA) based on the use of its dechlorinated analogue, ochratoxin B (OTB), in a displacement immunoassay. OTB was immobilised on controlled-pore glass beads followed by the binding of anti-OTA antibody, with anti-IgG antibody peroxidase conjugate used as a label. In this manner, an original bio-sensing material was obtained. Upon incubation of this material with OTA, the toxin competes with OTB for the binding sites of the anti-OTA antibodies and releases the antibody-tagged peroxidase complex into the solution. Compared to classic competitive immunoassays, this newly developed displacement immunoassay presents a similar detection limit and assay time. Moreover, the detection, based on the activity of the horseradish peroxidase, is performed for the first time in situ using wine samples.
Product Description
Supelco
Ochratoxin B solution, ~10 μg/mL in acetonitrile, analytical standard