MilliporeSigma
  • Fast and facile dissolution of cellulose with tetrabutylphosphonium hydroxide containing 40 wt% water.
Fast and facile dissolution of cellulose with tetrabutylphosphonium hydroxide containing 40 wt% water.
Chemical communications (Cambridge, England) (2012-01-06)
Mitsuru Abe, Yukinobu Fukaya, Hiroyuki Ohno
ABSTRACT
Tetrabutylphosphonium hydroxide containing 40 wt% water dissolved 20 wt% cellulose at the final concentration within 5 minutes under mild stirring at 25 °C.
Sigma-Aldrich
Tetrabutylphosphonium chloride, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrabutylphosphonium bromide, 98%
Pricing and availability is not currently available.