MilliporeSigma
  • Changes in intramuscular fat, fatty acid profile and cholesterol content induced by zeranol implantation strategy in hair lambs.
Changes in intramuscular fat, fatty acid profile and cholesterol content induced by zeranol implantation strategy in hair lambs.
Journal of the science of food and agriculture (2011-12-07)
Nidia V Valenzuela-Grijalva, Humberto González-Rios, Thalia Y Islava, Martin Valenzuela, Gastón Torrescano, Juan P Camou, Francisco A Núñez-González
ABSTRACT
The effect of zeranol implantation strategy on intramuscular fat, fatty acid profile and cholesterol content of the longissimus dorsi muscle of hair lambs was studied. Four treatments were tested: C, control group; Z12, 12 mg zeranol; Z24, 24 mg zeranol in a single application; and RZ12, 12 mg zeranol given twice. One-way analysis of variance was employed to estimate the effect of treatments (P < 0.05). To separate the effect of the mean, orthogonal contrasts were tested: C1, C versus Z12 + Z24 + RZ12; C2, Z12 versus Z24 + RZ12; and C3, Z24 versus RZ12. A decrease (P < 0.05) in intramuscular fat content was observed from implanting (C1 effect) and zeranol reimplantation (C3 effect). Implanted lambs exhibited an increase (P < 0.05) in monounsaturated fatty acids compared with control group (40.60% versus 35.35%). All contrasts were significant for the sum of n-6 and n-3, with values lower (P < 0.05) in the control (n-6: 0.84% and n-3: 1.38%) and higher in the RZ12 treatment (n-6: 7.55% and n-3: 14.9%). Cholesterol decreased by 78% with implantation and increasing the dose. The results indicate that it is possible to induce favorable changes in the fatty acid profile and cholesterol content using a zeranol implantation strategy on hair lambs.
Sigma-Aldrich
α-Zearalanol, ~98% (HPLC)