MilliporeSigma
  • Effects of vanillin and o-vanillin on induction of DNA-repair networks: modulation of mutagenesis in Escherichia coli.
Effects of vanillin and o-vanillin on induction of DNA-repair networks: modulation of mutagenesis in Escherichia coli.
Mutation research (1990-06-01)
K Takahashi, M Sekiguchi, Y Kawazoe
ABSTRACT
Vanillin and its isomer o-vanillin have an effect on the adaptive and SOS responses, as well as mutagenesis, induced in Escherichia coli by N-methyl-N-nitrosourea (MNU) and UV irradiation, potentiating in some cases and suppressing in others. o-Vanillin markedly inhibited the MNU-induced adaptive response, while both vanillins potentiated the UV-induced SOS response. These phenomena appear to be responsible for the comutagenic or antimutagenic role of these chemicals in MNU and UV mutagenesis.
Sigma-Aldrich
o-Vanillin, 99%