MilliporeSigma
  • Preparation of high purity biphenyl cyclooctene lignans from Schisandra extract by ion exchange resin catalytic transformation combined with macroporous resin separation.
Preparation of high purity biphenyl cyclooctene lignans from Schisandra extract by ion exchange resin catalytic transformation combined with macroporous resin separation.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-10-04)
Chun-hui Ma, Ting-ting Liu, Lei Yang, Yuan-gang Zu, Feng-jian Yang, Chun-jian Zhao, Lin Zhang, Zhong-hua Zhang
ABSTRACT
In this study, ester-bond biphenyl cyclooctene lignans were efficiently hydrolytically degraded into free biphenyl cyclooctene lignans by ion exchange resin transformation and simultaneous removal of impurities by macroporous resin. The OH-type strongly basic anion exchange resin 201×7 was the best one, and the dynamic hydrolysis efficiency was 146.7±5.0%. HPD5000 macroporous resin, which offered higher adsorption and desorption capacities and faster adsorption than other resins. The purity of free biphenyl cyclooctene lignans in the product increased from 5.14±0.24% to 79.67±0.0.67%. After dynamic catalytic transformation by 201×7 resin combined with purification of HPD5000 resin, the yield and the purity of free biphenyl cyclooctene lignans in the product were 132.1±4.7% and 80.91±3.53%, respectively.
Sigma-Aldrich
Schizandrin, ≥98% (HPLC)
Pricing and availability is not currently available.