MilliporeSigma
  • Discontinued and alternative brominated flame retardants in the atmosphere and precipitation from the great lakes basin.
Discontinued and alternative brominated flame retardants in the atmosphere and precipitation from the great lakes basin.
Environmental science & technology (2011-09-29)
Amina Salamova, Ronald A Hites
ABSTRACT
Air (vapor and particle) and precipitation samples were collected at five sites (two urban, one rural, and two remote) around the Great Lakes during 2005-2009 as a part of the Integrated Atmospheric Deposition Network (IADN). The concentrations of polybrominated diphenyl ethers (PBDEs), decabromodiphenylethane (DBDPE), hexabromobenzene (HBB), pentabromoethylbenzene (PBEB), and 1,2-bis(2,4,6-tribromophenoxy)ethane (BTBPE) were measured in these samples. The highest concentrations of these compounds were generally observed at the two urban sites-Chicago and Cleveland-with a few exceptions: The remote site at Eagle Harbor had particularly high levels of PBEB in all three phases, and the rural Sturgeon Point site had the highest HBB concentrations in the vapor phase. The sources of HBB and PBEB to these sites are unknown. A multiple linear regression model was applied to the concentrations of these compounds in the vapor phase, particle phase, precipitation, and the three phases combined. This regression resulted in overall (three phases combined) halving times for total PBDE concentrations of 6.3 ± 1.1 years. The overall halving times for HBB and BTBPE were 9.5 ± 4.6 years and 9.8 ± 2.8 years, respectively. For PBEB and DBDPE, the regression was not statistically significant for the combined phases, indicating that the atmospheric concentrations of these compounds have not changed between 2005 and 2009.
Sigma-Aldrich
Hexabromobenzene, 98%