MilliporeSigma
  • Exploring the metal coordination properties of the pyrimidine part of purine nucleobases: isomerization reactions in heteronuclear Pt(II)/Pd(II) of 9-methyladenine.
Exploring the metal coordination properties of the pyrimidine part of purine nucleobases: isomerization reactions in heteronuclear Pt(II)/Pd(II) of 9-methyladenine.
Inorganic chemistry (2011-09-24)
Susana Ibáñez, Francisca M Albertí, Pablo J Sanz Miguel, Bernhard Lippert
ABSTRACT
The synthesis and characterization of three heteronuclear Pt(2)Pd(2) (4, 5) and PtPd(2) (6) complexes of the model nucleobase 9-methyladenine (9-MeA) is reported. The compounds were prepared by reacting [Pt(NH(3))(3)(9-MeA-N7)](ClO(4))(2) (1) with [Pd(en)(H(2)O)(2)](ClO(4))(2) at different ratios r between Pt and Pd, with the goal to probe Pd(II) binding to any of the three available nitrogen atoms, N1, N3, N6 or combinations thereof. Pd(II) coordination occurs at N1 and at the deprotonated N6 positions, yet not at N3. 4 and 5 are isomers of [{(en)Pd}(2){N1,N6-9-MeA(-)-N7)Pt(NH(3))(3)}(2)](ClO(4))(6)·nH(2)O, with a head-head orientation of the two bridging 9-MeA(-) ligands in 4 and a head-tail orientation in 5. 6 is [{(en)Pd}(2)(OH)(N1,N6-9MeA(-)-N7)Pt(NH(3))(3)](ClO(4))(4)·4H(2)O, hence a condensation product between [Pt(NH(3))(3)(9-MeA-N7)](2+) and a μ-OH bridged dinuclear (en)Pd-OH-Pd(en) unit, which connects the N1 and N6 positions of 9-MeA(-) in an intramolecular fashion. 4 and 5, which slowly interconvert in aqueous solution, display distinct structural differences such as significantly different intramolecular Pd···Pd contacts (3.124 0(16) Å in 4; 2.986 6(14) Å in 5), among others. Binding of (en)Pd(II) to the exocyclic N6 atom in 4 and 5 is accompanied by a large movement of Pd(II) out of the 9-MeA(-) plane and a trend to a further shortening of the C6-N6 bond as compared to free 9-MeA. The packing patterns of 4 and 5 reveal substantial anion-π interactions.
Sigma-Aldrich
9-Methyladenine, 97%