MilliporeSigma
Degradation of 2-chlorotoluene by Rhodococcus sp. OCT 10.
Applied microbiology and biotechnology (2011-08-27)
Daniel Dobslaw, Karl-Heinrich Engesser
ABSTRACT
A strain Rhodococcus sp. OCT 10 DSM 45596(T), exhibiting 99.9% of 16S rDNA identity with Rhodococcus wratislaviensis NCIMB 13082, was isolated from a soil sample. The strain completely mineralised 2-chlorotoluene, 2-bromotoluene, o-xylene, benzyl alcohol and benzoate. In contrast, 2-fluorotoluene was only partially mineralised. By GC-MS and (1)H-NMR analyses, 4-chloro-3-methylcatechol was identified as the central intermediate in the degradation pathway of 2-chlorotoluene. It was further degraded by enzymes of the meta cleavage pathway. Catechol 1,2-dioxygenase and chlorocatechol 1,2-dioxygenase as the initial enzymes of the ortho cleavage pathways were not detectable under these conditions. Furthermore, neither formation nor oxidation of 2-chlorobenzylic alcohol, 2-chlorobenzaldehyde, or 2-chlorobenzoate was observed, thereby excluding side chain oxidation activity.
Sigma-Aldrich
2-Chlorotoluene, 99%