MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Direct and continuous fluorescence-based measurements of Pyrococcus horikoshii DNA N-6 adenine methyltransferase activity.
Direct and continuous fluorescence-based measurements of Pyrococcus horikoshii DNA N-6 adenine methyltransferase activity.
Analytical biochemistry (2011-08-16)
Michael D Maynard-Smith, Jennifer C McKelvie, Robert J Wood, Jenny E Harmer, Rohan T Ranasinghe, Claire L Williams, Danielle M Coomber, Alexandra F Stares, Peter L Roach
ABSTRACT
N-6 methylation of adenine destabilises duplex DNA and this can increase the proportion of DNA that dissociates into single strands. We have investigated utilising this property to measure the DNA adenine methyltransferase-catalyzed conversion of hemimethylated to fully methylated DNA through a simple, direct, fluorescence-based assay. The effects of methylation on the kinetics and thermodynamics of hybridisation were measured by comparing a fully methylated oligonucleotide product and a hemimethylated oligonucleotide substrate using a 13-bp duplex labeled on adjacent strands with a fluorophore (fluorescein) and quencher (dabcyl). Enzymatic methylation of the hemimethylated GATC site resulted in destabilisation of the duplex, increasing the proportion of dissociated DNA, and producing an observable increase in fluorescence. The assay provides a direct measurement of methylation rate in real time and is highly reproducible, with a coefficient of variance over 48 independent measurements of 3.6%. DNA methylation rates can be measured as low as 3.55 ± 1.84 fmols(-1) in a 96-well plate format, and the assay has been used to kinetically characterise the Pyrococcus horikoshii DNA adenine methyltransferase.
Product Description
Supelco
Butter Yellow, analytical standard