MilliporeSigma
Enantioselective synthesis of the C10-C20 fragment of fusicoccin A.
The Journal of organic chemistry (2011-07-16)
Anja Richter, Christian Hedberg, Herbert Waldmann
ABSTRACT
A synthesis of the fully protected C-ring fragment of the tricyclic diterpene fusicoccin A is reported. The desired cyclopentenyl halides 5a,b are obtained in a total of nine steps. Key transformations of the synthesis sequence are a nonconventional Cr-catalyzed allylic oxidation of a protected intermediate cylcopentenone, a diastereoselective addition of a propenyl Grignard/CeCl(3) reagent to the unmasked cyclopentenone, and an asymmetric hydroboration of the isopropenyl substituent. The protected and suitably functionalized C-ring fragment paves the way to explore further the total synthesis of fusicoccin A.
Sigma-Aldrich
Fusicoccin from Fusicoccum amygdali, ≥85% (HPLC)