MilliporeSigma
  • Comparison of 10-mg doses of 4-aminopyridine and 3,4-diaminopyridine for the treatment of downbeat nystagmus.
Comparison of 10-mg doses of 4-aminopyridine and 3,4-diaminopyridine for the treatment of downbeat nystagmus.
Journal of neuro-ophthalmology : the official journal of the North American Neuro-Ophthalmology Society (2011-07-08)
Roger Kalla, Rainer Spiegel, Jens Claassen, Stanislavs Bardins, Ales Hahn, Erich Schneider, Nicole Rettinger, Stefan Glasauer, Thomas Brandt, Michael Strupp
ABSTRACT
Animal experiments have demonstrated that aminopyridines increase Purkinje cell excitability, and in clinical studies, 4-aminopyridine (4-AP) and 3,4-diaminopyridine (3,4-DAP) improved downbeat nystagmus. In this double-blind, prospective, crossover study, the effects of equivalent doses of 4-AP and 3,4-DAP on the slow-phase velocity (SPV) of downbeat nystagmus were compared. Eight patients with downbeat nystagmus due to different etiologies (cerebellar degeneration [n = 1], bilateral vestibulopathy [n = 1], bilateral vestibulopathy and cerebellar degeneration [n = 1], Arnold-Chiari I malformation and cerebellar ataxia [n = 1], cryptogenic cerebellar ataxia [n = 4]) were included. They were randomly assigned to receiving a single capsule of 10 mg of 3,4-DAP or 4-AP followed by 6 days with no medication. One week later, the treatment was switched, that is, 1 single capsule (10 mg) of the other agent. Recordings with 3-dimensional video-oculography were performed before and 45 and 90 minutes after drug administration. Both medications had a significant effect throughout time (pre vs post 45 vs post 90) (F() = 8.876; P < 0.01). Following the administration of 3,4-DAP, mean slow velocity decreased from -5.68°/s (pre) to -3.29°/s (post 45) to -2.96°/s (post 90) (pre vs post 45/post 90 P < 0.01). In 4-AP, the mean SPV decreased from -6.04°/s (pre) to -1.58°/s (post 45) to -1.21°/s (post 90) (pre vs post 45/post 90 P < 0.00001). Both after 45 and after 90, the mean SPVs were significantly lower for 4-AP than for 3,4-DAP (P < 0.05). None of the patients reported serious side effects. Based on these results, 10-mg doses of 4-AP lead to a more pronounced decrease of the SPV of downbeat nystagmus than do equivalent doses of 3,4-DAP.
Sigma-Aldrich
3,4-Diaminopyridine, ≥98%