MilliporeSigma
  • Synthesis of unsymmetrical sulfides using ethyl potassium xanthogenate and recyclable copper catalyst under ligand-free conditions.
Synthesis of unsymmetrical sulfides using ethyl potassium xanthogenate and recyclable copper catalyst under ligand-free conditions.
The Journal of organic chemistry (2011-07-08)
Vijay Kumar Akkilagunta, Rama Rao Kakulapati
ABSTRACT
The synthesis of unsymmetrical sulfides has been achieved in good to excellent yields with inexpensive ethyl potassium xanthogenate via cross-coupling reaction using recyclable CuO nanoparticles under ligand-free conditions.The copper oxide nanoparticles can be recovered and reused up to five cycles without loss of activity.
Sigma-Aldrich
Potassium ethyl xanthogenate, 96%