Merck
  • Rare sugar D-allose suppresses gibberellin signaling through hexokinase-dependent pathway in Oryza sativa L.
Rare sugar D-allose suppresses gibberellin signaling through hexokinase-dependent pathway in Oryza sativa L.
Planta (2011-07-01)
Takeshi Fukumoto, Akihito Kano, Kouhei Ohtani, Yumiko Yamasaki-Kokudo, Bong-Gyu Kim, Kouji Hosotani, Miu Saito, Chikage Shirakawa, Shigeyuki Tajima, Ken Izumori, Toshiaki Ohara, Yoshio Shigematsu, Keiji Tanaka, Yutaka Ishida, Yoko Nishizawa, Yasuomi Tada, Kazuya Ichimura, Kenji Gomi, Kazuya Akimitsu
ABSTRACT
One of the rare sugars, D-allose, which is the epimer of D-glucose at C3, has an inhibitory effect on rice growth, but the molecular mechanisms of the growth inhibition by D-allose were unknown. The growth inhibition caused by D-allose was prevented by treatment with hexokinase inhibitors, D-mannoheptulose and N-acetyl-D-glucosamine. Furthermore, the Arabidopsis glucose-insensitive2 (gin2) mutant, which is a loss-of-function mutant of the glucose sensor AtHXK1, showed a D-allose-insensitive phenotype. D-Allose strongly inhibited the gibberellin-dependent responses such as elongation of the second leaf sheath and induction of α-amylase in embryo-less half rice seeds. The growth of the slender rice1 (slr1) mutant, which exhibits a constitutive gibberellin-responsive phenotype, was also inhibited by D-allose, and the growth inhibition of the slr1 mutant by D-allose was also prevented by D-mannoheptulose treatment. The expressions of gibberellin-responsive genes were down-regulated by D-allose treatment, and the down-regulations of gibberellin-responsive genes were also prevented by D-mannoheptulose treatment. These findings reveal that D-allose inhibits the gibberellin-signaling through a hexokinase-dependent pathway.
Sigma-Aldrich
D-Allose, 98%