MilliporeSigma
  • Linear PEI nanoparticles: efficient pDNA/siRNA carriers in vitro and in vivo.
Linear PEI nanoparticles: efficient pDNA/siRNA carriers in vitro and in vivo.
Nanomedicine : nanotechnology, biology, and medicine (2011-06-28)
Ritu Goyal, Sushil K Tripathi, Shilpa Tyagi, Anurag Sharma, K Ravi Ram, Debapratim K Chowdhuri, Yogeshwar Shukla, P Kumar, Kailash C Gupta
ABSTRACT
Linear polyethylenimine (lPEI, 25 kDa) nanoparticles' (LPN) series was synthesized by varying percentage of cross-linking with 1,4-butanediol diglycidyl ether (BDE) and their size, surface charge, morphology, pDNA protection/release, cytotoxicity and transfection efficiency were evaluated. Synthesized nanoparticles (NPs) were spherical in shape (size: ∼109 - 235 nm; zeta potential: +38 to +16 mV). These NPs showed increased buffering capacity with increasing percent cross-linking and also exhibited excellent transfection efficiency (i.e., ∼1.3 - 14.7 folds in case of LPN-5) in comparison with lPEI and the commercial transfection agents used in this study. LPN-5 based GFP-specific siRNA delivery resulted in ∼86% suppression of targeted gene expression. These particles were relatively nontoxic in vitro (in cell lines) and in vivo (in Drosophila). In vivo gene expression studies using LPN-5 in Balb/c mice through intravenous injection showed maximum expression of the reporter gene in the spleen. These results together demonstrate the potential of these particles as efficient transfection reagents. The authors demonstrate a novel method of synthesizing linear PEI nanoparticles to utilize these as transfection agents.
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, technical grade, 60%
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, ≥95%