MilliporeSigma
  • Optical band gap and spectroscopic study of lithium alumino silicate glass containing Y3+ ions.
Optical band gap and spectroscopic study of lithium alumino silicate glass containing Y3+ ions.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-22)
M S Shakeri, M Rezvani
ABSTRACT
The effect of different amounts of Y2O3 dopant on lithium alumino silicate (LAS) glass has been studied in this work. Glasses having 14.8Li2O-20Al2O3-65.2SiO2 (wt%) composition accompanied with Y2O3 dopant were prepared by normal melting process. In order to calculate the absorption coefficient of samples, transmittance and reflectance spectra of polished samples were measured in the room temperature. Optical properties i.e. Fermi energy level, direct and indirect optical band gaps and Urbach energy were calculated using functionality of extinction coefficient from Fermi-Dirac distribution function, Tauc's plot and the exponential part of absorption coefficient diagram, respectively. It has been clarified that variation in mentioned optical parameters is associated with the changes in physical properties of samples i.e. density or molar mass. On the other hand, increasing of Y3+ ions in the glassy microstructure of samples provides a semiconducting character to LAS glass by reducing the direct and indirect optical band gaps of glass samples from 1.97 to 1.67 and 3.46 to 2.1 (eV), respectively. These changes could be attributed to the role of Y3+ ions as the network former in the track of SiO4 tetrahedrals.
Sigma-Aldrich
Drying pearls orange
Pricing and availability is not currently available.