MilliporeSigma
  • Orientation of GST-tagged lectins via in situ surface modification to create an expanded lectin microarray for glycomic analysis.
Orientation of GST-tagged lectins via in situ surface modification to create an expanded lectin microarray for glycomic analysis.
Molecular bioSystems (2011-05-21)
Daniel C Propheter, Lara K Mahal
ABSTRACT
Herein we describe the orientation of GST-tagged lectins on NHS-activated slides via a one-step deposition of the protein and a glutathione (GSH) scaffold. This technology overcomes the need for a premade GSH-surface to orient GST-tagged proteins, enabling us to rapidly expand the analytical capacity of lectin microarrays through addition of oriented lectins, while maintaining lectin diversity.
Sigma-Aldrich
8-(6-Aminohexyl)aminoadenosine 3′:5′-cyclic monophosphate, ~98%