MilliporeSigma
  • Structural characterization of Y1 and Y2 receptors for neuropeptide Y and peptide YY by affinity cross-linking.
Structural characterization of Y1 and Y2 receptors for neuropeptide Y and peptide YY by affinity cross-linking.
The Journal of biological chemistry (1990-05-15)
S P Sheikh, J A Williams
ABSTRACT
Pharmacological studies indicate that peptide YY (PYY) and neuropeptide Y interact with multiple binding sites, categorized as Y1 and Y2 subtypes. In order to identify and structurally characterize the Y1 and Y2 receptors we covalently cross-linked [125I-Tyr36]PYY to its receptors. The Y2 receptor in rat hippocampus and rabbit kidney membranes was affinity labeled using different homo- and heterobifunctional cross-linking reagents. Analysis by sodium dodecyl sulfate-polyacrylamide gel electrophoresis and autoradiography resulted in a major labeled protein band of Mr = 50,000 in both hippocampal and kidney membranes, which was unaffected by reducing agents. The Y1 receptor was analyzed in membranes from the MC-IXC human neuroblastoma cell line. Autoradiography revealed two labeled bands at Mr = 70,000 and 45,000. As the intensity of the Mr = 45,000 band was reduced by protease inhibitors, it is likely that this band is a degradation product of the larger band. Labeling of these proteins was obtained only when N-5-azido-2-nitrobenzoyloxysuccinimide was employed for cross-linking followed by exposure to UV light. Labeling of the two cross-linked bands was unaffected by reducing agents. The binding of radiolabeled PYY and the intensity of the cross-linked bands, for both the Y1 and Y2 receptors, were inhibited similarly in a dose-dependent manner by increasing concentrations of unlabeled PYY. When exposed to agarose-coupled lectins, the detergent-solubilized Y1 receptor-hormone complex was completely adsorbed by wheat germ agglutinin and partially by ricin communis II. The cross-linked Y2 receptor was almost totally adsorbed by wheat germ agglutinin-agarose and partially adsorbed by concanavalin A. The adsorptions were in all cases blocked by the appropriate hapten sugar. These results indicate that the Y1 receptor is a glycoprotein with a Mr = 70,000 binding subunit, whereas the Y2 receptor is a glycoprotein with a Mr = 50,000 binding subunit. These results provide evidence that the Y1 and Y2 subtypes of neuropeptide Y and PYY receptors, previously characterized pharmacologically, are structurally distinct glycoproteins, not disulfide-linked to other subunits.