MilliporeSigma
  • Rationally engineered polymeric cisplatin nanoparticles for improved antitumor efficacy.
Rationally engineered polymeric cisplatin nanoparticles for improved antitumor efficacy.
Nanotechnology (2011-05-18)
Abhimanyu Paraskar, Shivani Soni, Sudipta Basu, Chitra J Amarasiriwardena, Nicola Lupoli, Shyam Srivats, Rituparna Sinha Roy, Shiladitya Sengupta
ABSTRACT
The use of cisplatin, a first line chemotherapy for most cancers, is dose-limited due to nephrotoxicity. While this toxicity can be addressed through nanotechnology, previous attempts at engineering cisplatin nanoparticles have been limited by the impact on the potency of cisplatin. Here we report the rational engineering of a novel cisplatin nanoparticle by harnessing a novel polyethylene glycol-functionalized poly-isobutylene-maleic acid (PEG-PIMA) copolymer, which can complex with cis-platinum (II) through a monocarboxylato and a coordinate bond. We show that this complex self-assembles into a nanoparticle, and exhibits an IC(50) = 0.77 ± 0.11 µM comparable to that of free cisplatin (IC(50) = 0.44 ± 0.09 µM). The nanoparticles are internalized into the endolysosomal compartment of cancer cells, and release cisplatin in a pH-dependent manner. Furthermore, the nanoparticles exhibit significantly improved antitumor efficacy in a 4T1 breast cancer model in vivo, with limited nephrotoxicity, which can be explained by preferential biodistribution in the tumor with reduced kidney concentrations. Our results suggest that the PEG-PIMA-cisplatin nanoparticle can emerge as an attractive solution to the challenges in cisplatin chemotherapy.
Sigma-Aldrich
Poly(isobutylene-alt-maleic anhydride), average Mw ~6,000, 12-200 mesh (85%)
Sigma-Aldrich
Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)], average Mw ~60,000