MilliporeSigma
  • Systemic antibiotics for chronic rhinosinusitis without nasal polyps in adults.
Systemic antibiotics for chronic rhinosinusitis without nasal polyps in adults.
The Cochrane database of systematic reviews (2011-05-13)
Patorn Piromchai, Sanguansak Thanaviratananich, Malinee Laopaiboon
ABSTRACT
Chronic rhinosinusitis (CRS) is a common health problem which significantly affects quality of life. A wide range of medical and surgical therapies have been used to treat CRS. Both systemic and topical antibiotics are used with the aim of eliminating infection and inflammation, altering bacterial biofilm formation, reversing ostial occlusion and improving symptoms. Various groups of systemic antibiotics have been studied; clinical cure rates reported are inconsistent and range from 50% to 95%. To determine the effectiveness and adverse reactions associated with systemic antibiotic therapy for CRS in adults. We searched the Cochrane ENT Group Trials Register; CENTRAL (2010, Issue 2); PubMed; EMBASE; CINAHL; Web of Science; BIOSIS Previews; Cambridge Scientific Abstracts; ISRCTN and additional sources for published and unpublished trials. The date of the most recent search was 10 June 2010. Randomised controlled trials (RCTs) comparing systemic antibiotics with placebo for chronic rhinosinusitis in adults. Two authors extracted data independently, compared results and resolved disagreements by discussion. We assessed treatment effect by calculating the risk ratio (RR) and 95% confidence intervals (CI) of cure at a specific time point for each trial. We used mean difference (MD) and 95% CI for continuous variables (severity scores, duration of symptoms, etc.). We included one study involving 64 patients. We assessed the overall risk of bias in this study as high. The study reported that roxithromycin could reduce the mean response score of patients by 0.73 points on a 1 to 6-point scale (95% CI 0.32 to 1.14, P = 0.0005) compared to those on placebo at three months after the start of treatment. This study also used sinonasal outcome test-20 (SNOT-20) comparing between pre and post-treatment at six, 12 and 24 weeks. The mean change in SNOT-20 from baseline at 24 weeks in the roxithromycin group was not significantly more than in the placebo group, at 0.27 points (95% CI -0.24 to 0.78, P = 0.30) on a 0 to 5-point scale. There is limited evidence from one small study to support the use of systemic antibiotics for the curative treatment of chronic rhinosinusitis in adults. Further good quality trials, with large sample sizes, are needed to evaluate the use of antibiotics in chronic rhinosinusitis.
Product Description
Roxithromycin, European Pharmacopoeia (EP) Reference Standard