MilliporeSigma
  • Toward canonical ensemble distribution from self-guided Langevin dynamics simulation.
Toward canonical ensemble distribution from self-guided Langevin dynamics simulation.
The Journal of chemical physics (2011-04-12)
Xiongwu Wu, Bernard R Brooks
ABSTRACT
This work derives a quantitative description of the conformational distribution in self-guided Langevin dynamics (SGLD) simulations. SGLD simulations employ guiding forces calculated from local average momentums to enhance low-frequency motion. This enhancement in low-frequency motion dramatically accelerates conformational search efficiency, but also induces certain perturbations in conformational distribution. Through the local averaging, we separate properties of molecular systems into low-frequency and high-frequency portions. The guiding force effect on the conformational distribution is quantitatively described using these low-frequency and high-frequency properties. This quantitative relation provides a way to convert between a canonical ensemble and a self-guided ensemble. Using example systems, we demonstrated how to utilize the relation to obtain canonical ensemble properties and conformational distributions from SGLD simulations. This development makes SGLD not only an efficient approach for conformational searching, but also an accurate means for conformational sampling.
Sigma-Aldrich
Ala-Ala
Sigma-Aldrich
D-Ala-D-Ala