MilliporeSigma
  • Novel potential markers of nasopharyngeal carcinoma for diagnosis and therapy.
Novel potential markers of nasopharyngeal carcinoma for diagnosis and therapy.
Clinical biochemistry (2011-03-16)
Faqing Tang, Chunlei Xie, Damao Huang, Yuan Wu, Maomao Zeng, Lunchao Yi, Yongfang Wang, Wenhua Mei, Ya Cao, Lunquan Sun
ABSTRACT
To search for markers of nasopharyngeal carcinoma (NPC) for diagnosis. Using gas chromatography and mass spectrometry, we evaluated 51 serum metabolites in 49 NPC, 37 throat cancer patients and 40 healthy controls. High metabolites were selected and confirmed in NPC tissues. Sensitivity and specificity were appraised for 53 NPC diagnoses. Metabolic profiling revealed that kynurenine, N-acetylglucosaminylamine, N-acetylglucosamine and hydroxyphenylpyruvate increased in NPC patient sera. Their sensitivity and specificity were respectively 79% and 71%, 78% and 69%, 83% and 68%, 84% and 73% for NPC diagnosis. These increases were confirmed in NPC cells. Four metabolites gradually increased from stage I to stage III. After radiotherapy, four metabolites decreased gradually, and tended to a normal level, and were associated with rate of tumor reduction. The results reveal that kynurenine, N-acetylglucosaminylamine, N-acetylglucosamine and hydroxyphenylpyruvate are potentially markers of NPC for diagnosis and therapy.
Sigma-Aldrich
Phenylpyruvic acid, 98%
Sigma-Aldrich
Sodium phenylpyruvate, powder