MilliporeSigma
  • Structure and saccharification of rice straw pretreated with sulfur trioxide micro-thermal explosion collaborative dilutes alkali.
Structure and saccharification of rice straw pretreated with sulfur trioxide micro-thermal explosion collaborative dilutes alkali.
Bioresource technology (2011-03-12)
Ri-sheng Yao, Hua-jia Hu, Sheng-song Deng, Huai Wang, Hui-xia Zhu
ABSTRACT
In this paper, a sulfur trioxide collaborative dilutes alkali method has been developed to pre-treat rice straw and it has been studied that the pre-treated rice straw structure affected the saccharification of the rice straw hydrolyzed by cellulose enzymatic hydrolysis. The results show that the reaction of the sulfur trioxide with rice straw resulted in the internal micro-thermal explosion, and the saccharification rate was 91% based on the pretreated rice straw with sulfur trioxide for 4h following 1% w/v NaOH treatment for 7h at 50°C.