MilliporeSigma
  • Dicetyl phosphate-tetraethylenepentamine-based liposomes for systemic siRNA delivery.
Dicetyl phosphate-tetraethylenepentamine-based liposomes for systemic siRNA delivery.
Bioconjugate chemistry (2011-03-03)
Tomohiro Asai, Saori Matsushita, Eriya Kenjo, Takuma Tsuzuku, Norihito Yonenaga, Hiroyuki Koide, Kentaro Hatanaka, Takehisa Dewa, Mamoru Nango, Noriyuki Maeda, Hiroshi Kikuchi, Naoto Oku
ABSTRACT
Dicetyl phosphate-tetraethylenepentamine (DCP-TEPA) conjugate was newly synthesized and formed into liposomes for efficient siRNA delivery. Formulation of DCP-TEPA-based polycation liposomes (TEPA-PCL) complexed with siRNA was examined by performing knockdown experiments using stable EGFP-transfected HT1080 human fibrosarcoma cells and siRNA for GFP. An adequate amount of DCP-TEPA in TEPA-PCL and N/P ratio of TEPA-PCL/siRNA complexes were determined based on the knockdown efficiency. Then, the biodistribution of TEPA-PCL modified with poly(ethylene glycol) (PEG) was examined in BALB/c mice. As a result, TEPA-PCL modified with PEG6000 avoided reticuloendothelial system uptake and showed long circulation in the bloodstream. On the other hand, PEGylation of TEPA-PCL/siRNA complexes caused dissociation of a portion of the siRNA from the liposomes. However, we found that the use of cholesterol-conjugated siRNA improved the interaction between TEPA-PCL and siRNA, which allowed PEGylation of TEPA-PCL/siRNA complexes without siRNA dissociation. In addition, TEPA-PCL complexed with cholesterol-conjugated siRNA showed potent knockdown efficiency in stable luciferase-transfected B16-F10 murine melanoma cells. Finally, the biodistribution of cholesterol-conjugated siRNA formulated in PEGylated TEPA-PCL was examined by performing near-infrared fluorescence imaging in Colon26 NL-17 murine carcinoma-bearing mice. Our results showed that tumor targeting with siRNA via systemic administration was achieved by using PEGylated TEPA-PCL combined with active targeting with Ala-Pro-Arg-Pro-Gly, a peptide used for targeting angiogenic endothelium.
Sigma-Aldrich
Dihexadecyl phosphate
Sigma-Aldrich
Tetraethylenepentamine, technical grade