MilliporeSigma
  • Electrospray ionization-ion mobility spectrometry as a detection system for three-phase hollow fiber microextraction technique and simultaneous determination of trimipramine and desipramine in urine and plasma samples.
Electrospray ionization-ion mobility spectrometry as a detection system for three-phase hollow fiber microextraction technique and simultaneous determination of trimipramine and desipramine in urine and plasma samples.
Analytical and bioanalytical chemistry (2011-02-08)
M T Jafari, M Saraji, H Sherafatmand
ABSTRACT
A novel method based on three-phase hollow fiber microextraction technique (HF-LPME) coupled with electrospray ionization-ion mobility spectrometry (ESI-IMS) was developed for the simultaneous determination of two antidepressant drugs (trimipramine and desipramine) in urine and plasma samples. The effects of various parameters such as type of organic solvent, composition of donor and acceptor phase, stirring rate, salt addition, extraction time, and temperature were investigated. Under the optimized conditions, the relative standard deviation was in the range of 5-6%, and the method quantitation limit (MQL) of utilizing HF-LPME/ESI-IMS was 5 μg/L for both drugs. The relative recoveries obtained by the proposed method from urine and plasma samples were in the range 94% to 97% for trimipramine and 92% to 96% for desipramine. Finally, the feasibility of the proposed method was successfully confirmed by extraction and determination of trace amounts of trimipramine and desipramine in biological samples without any significant matrix effect.
Sigma-Aldrich
Trimipramine maleate salt, ≥98% (TLC), powder