MilliporeSigma
Virtual screening for ligands of the insect molting hormone receptor.
Journal of chemical information and modeling (2011-02-01)
Toshiyuki Harada, Yoshiaki Nakagawa, Takehiko Ogura, Yutaka Yamada, Takehiro Ohe, Hisashi Miyagawa
ABSTRACT
Insect growth is regulated by the orchestrated event of ecdysteroids and their receptor proteins. Agonists/antagonists of ecdysteroid receptor are predicted to disrupt normal growth, providing good candidates of new insecticides. A database of over 2 million compounds was subjected to a shape-based virtual screening cascade to identify novel nonsteroidal hits similar to the known EcR ligand ponasterone A. Testing revealed micromolar hits against two strains of insect cells. Docking experiments against EcR were used to support the predicted binding mode of these ligands based on their overlay to ponasterone A.
Sigma-Aldrich
Ponasterone A, ≥65%