MilliporeSigma
  • Three-dimensional GaN/AlN nanowire heterostructures by separating nucleation and growth processes.
Three-dimensional GaN/AlN nanowire heterostructures by separating nucleation and growth processes.
Nano letters (2011-01-27)
Santino D Carnevale, Jing Yang, Patrick J Phillips, Michael J Mills, Roberto C Myers
ABSTRACT
Bottom-up nanostructure assembly has been a central theme of materials synthesis over the past few decades. Semiconductor quantum dots and nanowires provide additional degrees of freedom for charge confinement, strain engineering, and surface sensitivity-properties that are useful to a wide range of solid state optical and electronic technologies. A central challenge is to understand and manipulate nanostructure assembly to reproducibly generate emergent structures with the desired properties. However, progress is hampered due to the interdependence of nucleation and growth phenomena. Here we show that by dynamically adjusting the growth kinetics, it is possible to separate the nucleation and growth processes in spontaneously formed GaN nanowires using a two-step molecular beam epitaxy technique. First, a growth phase diagram for these nanowires is systematically developed, which allows for control of nanowire density over three orders of magnitude. Next, we show that by first nucleating nanowires at a low temperature and then growing them at a higher temperature, height and density can be independently selected while maintaining the target density over long growth times. GaN nanowires prepared using this two-step procedure are overgrown with three-dimensionally layered and topologically complex heterostructures of (GaN/AlN). By adjusting the growth temperature in the second growth step either vertical or coaxial nanowire superlattices can be formed. These results indicate that a two-step method allows access to a variety of kinetics at which nanowire nucleation and adatom mobility are adjustable.
Sigma-Aldrich
Aluminum nitride, nanopowder, <100 nm particle size
Sigma-Aldrich
Aluminum nitride, powder, 10 μm, ≥98%