MilliporeSigma
[Pharmacological correction of the cytogenetic effects of cisplatin].
Eksperimental'naia i klinicheskaia farmakologiia (2011-01-25)
O L Voronova, O V Neupokoeva, E P Fedorova, A E Prosenko, A A Churin
ABSTRACT
Beginning with the first hours of experiments, cisplatin evoked an increase of chromosomal aberrations in CBA/CaLac bone marrow cells. Significant increase of structural infringements of chromosomes due to chromatid breaks was revealed in metaphase plates of murine bone marrow preparations through 24 h after cisplatin intraperitoneal introduction. In late terms of research (90th day), the high level of aberrations of chromosomes was retained. The most pronounced correction of cisplatin mutagenicity was achieved using a preliminary course of thiophan introduction.
Sigma-Aldrich
Tetrahydrothiophene, 99%