MilliporeSigma
  • A new approach to 3-substituted tetrahydro-β-carboline derivative via diethyl acetamidomalonate.
A new approach to 3-substituted tetrahydro-β-carboline derivative via diethyl acetamidomalonate.
Amino acids (2010-11-10)
Sambasivarao Kotha, Shilpi Misra, Shaikh M Mobin
ABSTRACT
Strategically a new approach for the synthesis tetrahydro-β-carboline unit with the aid of diethyl acetamidomalonate as a glycine equivalent has been described.
Sigma-Aldrich
Diethyl acetamidomalonate-2-13C, ≥99 atom % 13C, ≥99% (CP)
Sigma-Aldrich
Diethyl acetamidomalonate, 98%