MilliporeSigma
  • 3,4-diaminopyridine for the treatment of Lambert-Eaton myasthenic syndrome.
3,4-diaminopyridine for the treatment of Lambert-Eaton myasthenic syndrome.
Expert review of clinical immunology (2010-10-29)
Paul W Wirtz, Maarten J Titulaer, Joop Ma van Gerven, Jan J Verschuuren
ABSTRACT
The Lambert-Eaton myasthenic syndrome (LEMS) is an autoimmune disease in which antibodies against voltage-gated calcium channels inhibit cholinergic neurotransmission. LEMS is clinically characterized by muscle weakness and autonomic dysfunction. 3,4-diaminopyridine (3,4-DAP) blocks potassium channels in nerve terminals, resulting in an increase in acetylcholine release. This article describes the four randomized placebo-controlled trials of 3,4-DAP in patients with LEMS. All trials demonstrated a significant effect on muscle strength and compound muscle action potential amplitude. Furthermore, the safety and tolerability of 3,4-DAP are reviewed. The side effects of 3,4-DAP are generally mild and most frequently consist of paresthesias, but epileptic seizures and arrhythmias have been described in patients using high doses. Given the efficacy and safety of 3,4-DAP in LEMS, this drug is the mainstay for symptomatic treatment of LEMS.
Sigma-Aldrich
3,4-Diaminopyridine, ≥98%