MilliporeSigma
  • Biodegradation kinetics of the benzimidazole fungicide thiophanate-methyl by bacteria isolated from loamy sand soil.
Biodegradation kinetics of the benzimidazole fungicide thiophanate-methyl by bacteria isolated from loamy sand soil.
Biodegradation (2010-10-27)
Mariusz Cycoń, Marcin Wójcik, Zofia Piotrowska-Seget
ABSTRACT
Degradation of the fungicide thiophanate-methyl (TM) by Enterobacter sp. TDS-1 and Bacillus sp. TDS-2 isolated from sandy soil previously treated with TM was studied in mineral salt medium (MSM) and soil. Both strains were able to grow in MSM supplemented with TM (50 mg l(-1)) as the sole carbon source. Over a 16 days incubation period, 60 and 77% of the initial dose of TM were degraded by strains TDS-1 and TDS-2, respectively, and disappearance of TM was described by first-order kinetics. Medium supplementation with glucose markedly stimulated bacterial growth; while the final rate of TM degradation was reduced by 21 and 27% for strains TDS-1 and TDS-2, respectively as compared to medium with TM only. Moreover, this additional carbon source changed the TM degradation kinetics, which proceeded according to a zero-order model. This effect was linked to substrate competition and/or a strong decrease of medium pH. Isolates degraded TM (100 mg kg(-1)) in soil with rate constants of 0.186 and 0.210 day(-1), following first-order rate kinetics, and the time in which the initial TM concentration was reduced by 50% (DT50) in soils inoculated with strains TDS-1 and TDS-2 were 6.3 and 5.1 days, respectively. Analysis of TM degradation products in soil showed that the tested strains may have the potential to transform carbendazim (MBC) to 2-aminobenzimidazole (2-AB), and may be useful for a bioremediation of MBC-polluted soils.
Product Description
Supelco
Thiophanate-methyl, PESTANAL®, analytical standard
Pricing and availability is not currently available.