MilliporeSigma
  • Preparation and periodic emission of superlattice CdS/CdS:SnS2 microwires.
Preparation and periodic emission of superlattice CdS/CdS:SnS2 microwires.
Journal of the American Chemical Society (2010-08-19)
Guozhang Dai, Bingsuo Zou, Zhonglin Wang
ABSTRACT
Semiconductor superlattice micro-/nanowires could greatly increase the versatility and power of modulating electronic (or excitonic, photonic) transport, optical properties. In this communication, we report growth of a semiconductor CdS/CdS:SnS(2) superlattice microwire through a coevaporation technique with microenvironmental control. Such a novel superlattice microwire can modulate the exciton and photons to show multipeak emissions with periods in a wide spectral range, which arise in the 1-d photon crystal and confined exciton emission. This system can be widely used in producing multicolor emissions, low-threshold lasing, study light-matter interaction, slow light engineering, and weak nonlinear optical devices.