MilliporeSigma
  • Determination of formal potential of NADH/NAD+ redox couple and catalytic oxidation of NADH using poly(phenosafranin)-modified carbon electrodes.
Determination of formal potential of NADH/NAD+ redox couple and catalytic oxidation of NADH using poly(phenosafranin)-modified carbon electrodes.
Bioelectrochemistry (Amsterdam, Netherlands) (2010-07-30)
Farhana S Saleh, Mohammad R Rahman, Takeyoshi Okajima, Lanqun Mao, Takeo Ohsaka
ABSTRACT
The electrochemical regeneration of NADH/NAD(+) redox couple has been studied using poly(phenosafranin) (PPS)-modified carbon electrodes to evaluate the formal potential and catalytic rate constant for the oxidation of NADH. The PPS-modified electrodes were prepared by electropolymerization of phenosafranin onto different carbon substrates (glassy carbon (GC) and basal-plane pyrolytic graphite (BPPG)) in different electrolytic solutions. The formal potential was estimated to be -0.365±0.002V vs. SHE at pH 7.0. As for the bare carbon electrodes, the oxidation of NADH at the BPPG electrode was found to be enhanced compared with the GC electrode. For the PPS-modified electrodes, it was found that the electrocatalysis of PPS-modified electrodes for the oxidation of NADH largely depends on the carbon substrate and electrolyte solution employed for their preparation, i.e., the PPS-modified BPPG electrode prepared in 0.2M NaClO(4)/acetonitrile solution exhibits an excellent and persistent electrocatalytic property toward NADH oxidation in phosphate buffer solution (pH 7.0) with a diminution of the overpotential of about 740 and 670mV compared with those at the bare GC electrode and the PPS-modified GC electrode prepared in 0.2M H(2)SO(4) solution, respectively. A quantitative analysis of the electrocatalytic reaction based on rotating disk voltammetry gave the electrocatalytic reaction rate constants of the order of 10(3)-10(4)M(-)(1)s(-1) depending on the preparation conditions of the PPS-modified electrodes.
Sigma-Aldrich
Phenosafranin, Dye content 80 %