MilliporeSigma
  • Bufferized solvent extraction and HPLC fluorometric detection method for sarafloxacin in pig and chicken muscles.
Bufferized solvent extraction and HPLC fluorometric detection method for sarafloxacin in pig and chicken muscles.
Biomedical chromatography : BMC (2010-07-06)
Jeong-Heui Choi, Tae Woong Na, Md Iqbal Rouf Mamun, A M Abd El-Aty, Eun-Ho Shin, Jong-Hyouk Park, Jae-Han Shim
ABSTRACT
In this study, a method for the detection of sarafloxacin in pig and chicken muscles was developed using HPLC-FLD as a regulatory residue technique. Good extraction efficiency was achieved using a mixture of 1% orthophosphoric acid-0.2 m MgCl(2) in water and acetonitrile as an extraction solvent, and n-hexane partitioning and centrifugation for cleanup was used in the absence of dehydration. Specificity, linearity, detection and quantification limits, recovery, accuracy and precision were all validated, and all results were sufficient for the SARA regulatory residue method in pig and chicken muscles. The method developed and described herein was not only simple but also reliable, and was applied to market samples to determine their residue contents.
Product Description
Supelco
Sarafloxacin hydrochloride hydrate, VETRANAL®, analytical standard
Pricing and availability is not currently available.