MilliporeSigma
  • Efficient synthesis of 6-O-palmitoyl-1,2-O-isopropylidene-α-D-glucofuranose in an organic solvent system by lipase-catalyzed esterification.
Efficient synthesis of 6-O-palmitoyl-1,2-O-isopropylidene-α-D-glucofuranose in an organic solvent system by lipase-catalyzed esterification.
Biotechnology letters (2010-06-25)
Takashi Kobayashi, Taro Ehara, Takanori Mizuoka, Shuji Adachi
ABSTRACT
In order to synthesize a sugar ester at high concentration, 1,2-O-isopropylidene-α-D-glucofuranose (IpGlc), which is one of the sugar acetals and is more hydrophobic than unmodified glucose, was esterified with palmitic acid at 40°C using immobilized lipase from Candida antarctica in some organic solvents or their mixtures. Acetone + t-butyl alcohol (3:1 v/v) improved both the initial reaction rate and yield after 80 h: the product reached its maximum value (240 mmol/kg solvent; ca. 110 g/kg solvent) when 400 mmol IpGlc/kg solvent and 1,200 mmol palmitic acid/kg solvent were used in this solvent mixture.